accessibility menu, dialog, popup

UserName

H3COOCH3CH3HO

CAS RN 80-55-7

IUPAC Name: ethyl 2-hydroxy-2-methylpropanoate
Synonyms: ethyl 2-hydroxy-2-methylpropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: GFUIDHWFLMPAGY-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)