accessibility menu, dialog, popup

UserName

H3CCH3CH3H3CO

CAS RN 80-54-6

IUPAC Name: 3-(4-tert-butylphenyl)-2-methylpropanal
Synonyms: lilial
Molecular Weight (g/mol): 204.31
Molecular Formula: C14H20O
InChi Key: SDQFDHOLCGWZPU-UHFFFAOYNA-N
SMILES: CC(CC1=CC=C(C=C1)C(C)(C)C)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)