accessibility menu, dialog, popup

UserName

Uncover New Spotlight Deals and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 80-44-4

CH3OSOO

CAS RN 80-44-4

IUPAC Name: butyl benzenesulfonate
Synonyms: butyl benzenesulfonate
Molecular Weight (g/mol): 214.28
Molecular Formula: C10H14O3S
InChi Key: NIKBCKTWWPVAIC-UHFFFAOYSA-N
SMILES: CCCCOS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.