accessibility menu, dialog, popup

UserName

CAS RN 80-08-0

IUPAC Name: 4-(4-aminobenzenesulfonyl)aniline
Synonyms: dapsone N'-diphenyl sulfondiamide N dapson-fatol diaphenylsulfone dds sulphonyldianiline sulfone ucb sulfanona-mae 4,4'-sulfonylbisbenzeneamine dumitone P,P-sulphonylbisbenzamine
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: MQJKPEGWNLWLTK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (5)