accessibility menu, dialog, popup

UserName

FBrN

CAS RN 79630-23-2

IUPAC Name: 3-bromo-4-fluorobenzonitrile
Synonyms: 3-bromo-4-fluorobenzonitrile
Molecular Weight (g/mol): 200.01
Molecular Formula: C7H3BrFN
InChi Key: JKCYKISVUIVZCS-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Br)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)