accessibility menu, dialog, popup

UserName

ClCH3HN(S)(S)ClClH

CAS RN 79559-97-0

IUPAC Name: hydrogen (1S,4S)-4-(3,4-dichlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine chloride
Synonyms: hydrogen sertraline chloride
Molecular Weight (g/mol): 342.69
Molecular Formula: C17H18Cl3N
InChi Key: BLFQGGGGFNSJKA-XHXSRVRCSA-N
SMILES: [H+].[Cl-].CN[C@H]1CC[C@@H](C2=CC=C(Cl)C(Cl)=C2)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)