CAS RN 79538-29-7
IUPAC Name:
2,4,6-trifluorobenzoyl chloride
Synonyms:
2,4,6-trifluorobenzoyl chloride
Molecular Weight (g/mol):
194.54
Molecular Formula:
C7H2ClF3O
InChi Key:
SIFIJQFBERMWMU-UHFFFAOYSA-N
SMILES:
FC1=CC(F)=C(C(Cl)=O)C(F)=C1
1
–
1
of
1
results
1
2,4,6-Trifluorobenzoyl chloride, 97%
CAS: 79538-29-7 Molecular Formula: C7H2ClF3O Molecular Weight (g/mol): 194.537 MDL Number: MFCD00070810 InChI Key: SIFIJQFBERMWMU-UHFFFAOYSA-N Synonym: 2,4,6-trifluorobenzoylchloride,2,4,6-trifluoro-benzoyl chloride,acmc-20aof3,timtec-bb sbb005827,2,4,6-tri-fluorobenzoyl chloride,2,4,6-trifluorobenzoyl chloride,benzoyl chloride,2,4,6-trifluoro,benzoyl chloride, 2,4,6-trifluoro,2,4,6-trifluorobenzoyl chloride 1g,benzoyl chloride, 2,4,6-trifluoro-9ci PubChem CID: 144895 IUPAC Name: 2,4,6-trifluorobenzoyl chloride SMILES: C1=C(C=C(C(=C1F)C(=O)Cl)F)F