accessibility menu, dialog, popup

UserName

CAS RN 79-15-2

IUPAC Name: N-bromoacetamide
Synonyms: acetamide, N-bromo-
Molecular Weight (g/mol): 137.96
Molecular Formula: C2H4BrNO
InChi Key: VBTQNRFWXBXZQR-UHFFFAOYSA-N
SMILES: CC(=O)NBr
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results