CAS RN 78916-49-1
IUPAC Name:
1,3-dimethoxy-5-[(1E)-2-phenylethenyl]benzene
Synonyms:
1,3-dimethoxy-5-[(1E)-2-phenylethenyl]benzene
Molecular Weight (g/mol):
240.3
Molecular Formula:
C16H16O2
InChi Key:
BIYGTLDPTJMNET-CMDGGOBGSA-N
SMILES:
COC1=CC(\C=C\C2=CC=CC=C2)=CC(OC)=C1
1
–
1
of
1
results
1
3,5-Dimethoxystilbene 98.0+%, TCI America™
CAS: 78916-49-1 Molecular Formula: C16H16O2 Molecular Weight (g/mol): 240.30 MDL Number: MFCD00210545 InChI Key: BIYGTLDPTJMNET-CMDGGOBGSA-N PubChem CID: 5316874 IUPAC Name: 1,3-dimethoxy-5-[(1E)-2-phenylethenyl]benzene SMILES: COC1=CC(\C=C\C2=CC=CC=C2)=CC(OC)=C1