missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxystilbene 98.0+%, TCI America™

Catalog No. D25921G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25921G Supplier TCI America Supplier No. D25921G
Only null left

Chemical Identifiers

CAS 78916-49-1
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.30
MDL Number MFCD00210545
InChI Key BIYGTLDPTJMNET-CMDGGOBGSA-N
PubChem CID 5316874
IUPAC Name 1,3-dimethoxy-5-[(1E)-2-phenylethenyl]benzene
SMILES COC1=CC(\C=C\C2=CC=CC=C2)=CC(OC)=C1

Specifications

Melting Point 56°C
Color Yellow
Quantity 1 g
Formula Weight 240.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxystilbene