accessibility menu, dialog, popup

UserName

CAS RN 78842-13-4

IUPAC Name: 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 2-amino-9-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol): 285.24
Molecular Formula: C10H12FN5O4
InChi Key: UXUZARPLRQRNNX-SJLGBTOWNA-N
SMILES: NC1=NC2=C(N=CN2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2F)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results