accessibility menu, dialog, popup

UserName

CAS RN 78603-95-9

CH3CH3OHH2N

CAS RN 78603-95-9

IUPAC Name: 2-amino-3-methyl-1,1-diphenylbutan-1-ol
Synonyms: 2-amino-3-methyl-1,1-diphenylbutan-1-ol
Molecular Weight (g/mol): 255.36
Molecular Formula: C17H21NO
InChi Key: LNQVZZGGOZBOQS-UHFFFAOYNA-N
SMILES: CC(C)C(N)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results