CAS RN 78092-53-2
IUPAC Name:
5,11,17,23,29,35-hexa-tert-butylheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Synonyms:
5,11,17,23,29,35-hexa-tert-butylheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15,17,19(40),21,23,25(39),27,29,31(38),33,35-octadecaene-37,38,39,40,41,42-hexol
Molecular Weight (g/mol):
973.39
Molecular Formula:
C66H84O6
InChi Key:
UOEYZAXKBKAKRO-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(C2)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
1
–
1
of
1
results
1
Promotions Available
4-tert-Butylcalix[6]arene, 96%
4-tert-Butylcalix[6]arene, 96%
CAS: 78092-53-2 Molecular Formula: C66H84O6 Molecular Weight (g/mol): 973.392 MDL Number: MFCD00075464 InChI Key: UOEYZAXKBKAKRO-UHFFFAOYSA-N Synonym: 4-tert-butylcalix 6 arene,unii-b9og173dmy,b9og173dmy,5,11,17,23,29,35-hexa-tert-butylcalix 6 arene-37,38,39,40,41,42-hexol,hexa-tert-butyl hexahydroxy calix 6 arene,calx-b 6,p-t-butylcalix 6 arene,tert-butylcalix 6 arene,p-tert-butylcalix-6-arene PubChem CID: 335356 ChEBI: CHEBI:51251 SMILES: CC(C)(C)C1=CC2=C(C(=C1)CC3=CC(=CC(=C3O)CC4=CC(=CC(=C4O)CC5=CC(=CC(=C5O)CC6=C(C(=CC(=C6)C(C)(C)C)CC7=C(C(=CC(=C7)C(C)(C)C)C2)O)O)C(C)(C)C)C(C)(C)C)C(C)(C)C)O