CAS RN 780820-43-1
IUPAC Name:
3-(trimethylsilyl)naphthalen-2-yl trifluoromethanesulfonate
Synonyms:
3-(trimethylsilyl)naphthalen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol):
348.41
Molecular Formula:
C14H15F3O3SSi
InChi Key:
OSZYFPWWHQEAGW-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C1=C(OS(=O)(=O)C(F)(F)F)C=C2C=CC=CC2=C1
1
–
1
of
1
results
1
3-(Trimethylsilyl)-2-naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™
CAS: 780820-43-1 Molecular Formula: C14H15F3O3SSi Molecular Weight (g/mol): 348.411 InChI Key: OSZYFPWWHQEAGW-UHFFFAOYSA-N Synonym: 3-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 3-(Trimethylsilyl)-2-naphthyl Ester PubChem CID: 44629848 IUPAC Name: (3-trimethylsilylnaphthalen-2-yl) trifluoromethanesulfonate SMILES: C[Si](C)(C)C1=CC2=CC=CC=C2C=C1OS(=O)(=O)C(F)(F)F