accessibility menu, dialog, popup

UserName

CAS RN 78-94-4

IUPAC Name: but-3-en-2-one
Synonyms: methyl vinyl ketone methylvinylcetone 2-butenone delta(sup 3)-2-butenone methyl-vinyl-cetone delta(3)-2-butenone
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: FUSUHKVFWTUUBE-UHFFFAOYSA-N
SMILES: CC(=O)C=C
Boiling Point 
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)