accessibility menu, dialog, popup

UserName

NH4OSOOOZnOSOOONH4

CAS RN 77998-33-5

IUPAC Name: zinc(2+) diammonium disulfate
Synonyms: zinc(2+) diammonium disulfate
Molecular Weight (g/mol): 293.57
Molecular Formula: H8N2O8S2Zn
InChi Key: CHORECZHRAUKSW-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].[Zn++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results