accessibility menu, dialog, popup

UserName

CAS RN 7789-33-5

IUPAC Name: iodobromane
Synonyms: iodine monobromide
Molecular Weight (g/mol): 206.81
Molecular Formula: BrI
InChi Key: CBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILES: BrI
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)