missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98% min

Catalog No. AA3910622
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3910622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3910622 Supplier Thermo Scientific Chemicals Supplier No. 03910622

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7789-33-5 | BrI | 206.81 g/mol

Iodine bromide is used for bromination of and for determining iodine used. IBr is used in some types of iodometry. It is also used as a organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Boiling Point 116°C (decomposition)
Melting Point 50° (sublimation)
Physical Form Crystalline
Quantity 100 g
Flash Point None
Sensitivity Light and moisture sensitive
Assay Percent Range 98% min
Density 4.416 g/mL
Beilstein 3902973
Solubility Information It is soluble in alcohol,water,ether,carbon disulfi,and glacial acetic acid.
Formula Weight 206.81
Chemical Name or Material Iodine monobromide
Show More Show Less
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-159-9
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only