missing translation for 'onlineSavingsMsg'
Learn More

Iodobromide TS, Ricca Chemical

Catalog No. 410216
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
410216 500 mL
41024 120 mL
410232 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 410216 Supplier Ricca Chemical Company Supplier No. 410216
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Iodobromide TS
CAS 7789-33-5, 64-19-7
CAS Min % 96.0, 1.83
CAS Max % 100.0, 1.91
Molecular Formula BrI
MDL Number MFCD00011353
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILES BrI
IUPAC Name iodobromane
Molecular Weight (g/mol) 206.81
PubChem CID 82238
Grade Laboratory
Packaging Amber Glass Bottle
pH <1
Quantity 500 mL
Show More Show Less