accessibility menu, dialog, popup

UserName

CAS RN 7789-27-7

FTl

CAS RN 7789-27-7

IUPAC Name: λ¹-thallanylium fluoride
Synonyms: thallium(I) fluoride
Molecular Weight (g/mol): 223.38
Molecular Formula: FTl
InChi Key: CULOEOTWMUCRSJ-UHFFFAOYSA-M
SMILES: [F-].[Tl+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results