accessibility menu, dialog, popup

UserName

SrOCrOOO

CAS RN 7789-06-2

IUPAC Name: strontium(2+) dioxochromiumbis(olate)
Synonyms: strontium chromate
Molecular Weight (g/mol): 203.61
Molecular Formula: CrO4Sr
InChi Key: NVKTUNLPFJHLCG-UHFFFAOYSA-N
SMILES: [Sr++].[O-][Cr]([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results