CAS RN 7787-70-4
CAS RN 7787-70-4
IUPAC Name:
λ¹-copper(1+) bromide
Synonyms:
copper(I) bromide
Molecular Weight (g/mol):
143.45
Molecular Formula:
BrCu
InChi Key:
NKNDPYCGAZPOFS-UHFFFAOYSA-M
SMILES:
[Cu+].[Br-]
1
–
2
of
2
results
1
Copper(I) bromide, 98%, extra pure
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: bromocopper SMILES: [Cu+].[Br-]
2
Copper(I) bromide, 98%
CAS: 7787-70-4 Molecular Formula: BrCu Molecular Weight (g/mol): 143.45 MDL Number: MFCD00010969 InChI Key: NKNDPYCGAZPOFS-UHFFFAOYSA-M Synonym: copper i bromide,copper bromide cubr,copper 1+ bromide,copper monobromide,cuprousbromide,hsdb 270,cubr,copper 1 bromide PubChem CID: 24593 IUPAC Name: bromocopper SMILES: [Cu+].[Br-]