accessibility menu, dialog, popup

UserName

IIIBi

CAS RN 7787-64-6

IUPAC Name: bismuth(3+) triiodide
Synonyms: bismuth(III) iodide
Molecular Weight (g/mol): 589.69
Molecular Formula: BiI3
InChi Key: KOECRLKKXSXCPB-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Bi+3]
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results