accessibility menu, dialog, popup

UserName

BeH2OH2OH2OH2OOSOOO

CAS RN 7787-56-6

IUPAC Name: beryllium(2+) tetrahydrate sulfate
Synonyms: beryllium sulfate tetrahydrate beryllium sulfate, tetrahydrate (1:1:4)
Molecular Weight (g/mol): 177.13
Molecular Formula: BeH8O8S
InChi Key: DIMYTQPLZWDZFE-UHFFFAOYSA-L
SMILES: [Be++].O.O.O.O.[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results