accessibility menu, dialog, popup

UserName

CAS RN 7783-68-8

IUPAC Name: niobium(5+) pentafluoride
Synonyms: niobium(V) fluoride
Molecular Weight (g/mol): 187.9
Molecular Formula: F5Nb
InChi Key: AOLPZAHRYHXPLR-UHFFFAOYSA-I
SMILES: [F-].[F-].[F-].[F-].[F-].[Nb+5]
Grade 
  • (8)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results