CAS RN 7783-18-8
IUPAC Name:
dihydroxy-1λ⁶-disulfen-1-one diamine
Synonyms:
ammonium thiosulfate
Molecular Weight (g/mol):
148.2
Molecular Formula:
H8N2O3S2
InChi Key:
XYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILES:
N.N.OS(O)(=O)=S
1
–
3
of
3
results
1
Ammonium thiosulfate, 96%
CAS: 7783-18-8 Molecular Formula: (NH4)2S2O3 MDL Number: MFCD00015965 Synonym: ammonium thiosulphate,thiosulfuric acid h2s2o3 , diammonium salt,thiosulfuric acid h2s2o3 , ammonium salt 1:2,diammonium hypo,acmc-1bhnz,ammoniumthiosulfate,dsstox_cid_9703,dsstox_rid_78814,dsstox_gsid_29703,diammonium thiosulphate
2
Ammonium thiosulfate, 96%, pure
CAS: 7783-18-8 Molecular Formula: H8N2O3S2 Molecular Weight (g/mol): 148.21 MDL Number: MFCD00015965 Synonym: ammonium thiosulphate,thiosulfuric acid h2s2o3 , diammonium salt,thiosulfuric acid h2s2o3 , ammonium salt 1:2,diammonium hypo,acmc-1bhnz,ammoniumthiosulfate,dsstox_cid_9703,dsstox_rid_78814,dsstox_gsid_29703,diammonium thiosulphate
3