accessibility menu, dialog, popup

UserName

CAS RN 7781-98-8

IUPAC Name: ethyl 3-hydroxybenzoate
Synonyms: ethyl-3-hydroxybenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: MWSMNBYIEBRXAL-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(O)=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)