missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Hydroxybenzoate 99.0+%, TCI America™

Catalog No. E033325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E033325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E033325G Supplier TCI America Supplier No. E033325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7781-98-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002296
InChI Key MWSMNBYIEBRXAL-UHFFFAOYSA-N
Synonym ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate
PubChem CID 24522
IUPAC Name ethyl 3-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)O

Specifications

Melting Point 73°C
Boiling Point 167°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-Hydroxybenzoate