missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxybenzoate, 99%

Catalog No. AAA1534336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1534336 500 g
AAA1534314 25 g
AAA1534322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1534336 Supplier Thermo Scientific Chemicals Supplier No. A1534336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7781-98-8 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Ethyl 3-hydroxybenzoate has been used in the synthesis of chiral derivatives of ethyl 3-(2-methylbutyloxy)benzoate and 4-(2-methylbutyloxy)-4′-acetylbiphenyl, via Mitsunobu reaction. It is a food-grade antimicrobial agent.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7781-98-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002296
InChI Key MWSMNBYIEBRXAL-UHFFFAOYSA-N
Synonym ethyl-3-hydroxybenzoate, benzoic acid, 3-hydroxy-, ethyl ester, 3-hydroxybenzoic acid ethyl ester, m-ethoxycarbonylphenol, ethyl m-hydroxybenzoate, 3-hydroxy-benzoic acid ethyl ester, benzoic acid, m-hydroxy-, ethyl ester, pubchem19923, acmc-209pbw, ethyl 3-oxidanylbenzoate
PubChem CID 24522
IUPAC Name ethyl 3-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)O

Specifications

Melting Point 72°C to 76°C
Boiling Point 187°C to 188°C (31 mmHg)
Quantity 500 g
Beilstein 2208566
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Ethyl 3-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-951-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only