missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium dichromate, 0.1N Standardized Solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Packaging | Plastic bottle |
| Physical Form | Liquid |
| Linear Formula | K2Cr2O7 |
| Type | Analytical Volumetric Solution |
| Quantity | 1 L |
| Formula Weight | 294.18 |
| Grade | Standard |
| Chemical Name or Material | Potassium dichromate |
Safety and Handling
| Hazard Category | H317-H332-H334-H340-H350-H360FD |
| Precautionary Statement | P201-P202-P260-P271-P281-P304+P340-P308+P313-P501c |
| EINECSNumber | 231-906-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only