missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111018413
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018413 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018413 Supplier Merck Emd Millipore Supplier No. 4201950GF
Only null left

Potassium dichromate is a reference toxicant which may be used in many laboratories for aquatic toxicity testing. It can also find applications as an oxidant involved in the chemical oxidative polymerization to prepare conducting and electro-active polymers like polyaniline and polypyrrole.

Specifications

CAS 7778-50-9
Physical Form Neat
Linear Formula K2 Cr2O7
MDL Number MFCD00011367
Melting Point 398°C
Synonym Potassium bichromate
Type Certified Reference Material
Quantity 50 g
Formula Weight 294.18
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less