CAS RN 7778-18-9
Drierite™, with indicator, 8 mesh
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™ Drying Desiccants, W.A. Hammond
Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, with indicator, 10-20 mesh
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, indicating (CaSO{4})
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, indicating (CaSO{4})
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, indicating (CaSO{4})
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, regular (CaSO{4}), Lab Grade
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Calcium sulfate, anhydrous, 99%
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L Synonym: calcium sulfate,drierite,gypsum,sulfuric acid, calcium salt 1:1,karstenite,calcium sulphate,anhydrous gypsum,plaster of paris,anhydrous calcium sulfate,calcium sulfate, anhydrous PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium;sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, without indicator, 10-20 mesh
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, indicating (CaSO{4})
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, without indicator, 4 mesh
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 SMILES: [Ca++].[O-]S([O-])(=O)=O
DRIERITE™, Indicating, 10 to 20 Mesh, J.T. Baker™
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, without indicator, 20-40 mesh
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, regular (CaSO{4}), Lab Grade
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
Drierite™, regular (CaSO{4}), Lab Grade
CAS: 7778-18-9 Molecular Formula: CaO4S Molecular Weight (g/mol): 136.13 MDL Number: MFCD00010912 InChI Key: OSGAYBCDTDRGGQ-UHFFFAOYSA-L PubChem CID: 24497 ChEBI: CHEBI:31346 IUPAC Name: calcium sulfate SMILES: [Ca++].[O-]S([O-])(=O)=O
- 1
- 2