accessibility menu, dialog, popup

UserName

H3COOCH

CAS RN 77758-51-1

IUPAC Name: methyl hex-5-ynoate
Synonyms: methyl hex-5-ynoate
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: LZULAZTXJLWELL-UHFFFAOYSA-N
SMILES: COC(=O)CCCC#C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)