accessibility menu, dialog, popup

UserName

CAS RN 7748-36-9

IUPAC Name: oxetan-3-ol
Synonyms: oxetan-3-ol
Molecular Weight (g/mol): 74.08
Molecular Formula: C3H6O2
InChi Key: QMLWSAXEQSBAAQ-UHFFFAOYSA-N
SMILES: OC1COC1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)