accessibility menu, dialog, popup

UserName

CAS RN 7745-91-7

CH3NH2Br

CAS RN 7745-91-7

IUPAC Name: 3-bromo-4-methylaniline
Synonyms: 3-bromo-4-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: GRXMMIBZRMKADT-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)