missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylaniline 98.0+%, TCI America™

Catalog No. B23545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23545G 5 g
B235425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23545G Supplier TCI America Supplier No. B23545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7745-91-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00134176
InChI Key GRXMMIBZRMKADT-UHFFFAOYSA-N
Synonym 3-bromo-p-toluidine, p-toluidine, 3-bromo, benzenamine, 3-bromo-4-methyl, 3-bromo-4-methyl-phenylamine, 3-bromo-1,4-toluidine, 4-amino-2-bromotoluene, 3-bromo-4-methylbenzenamine, 3-bromo-4-methylphenylamine, 3-bromo-para-toluidine, 3-bromo-4-methyl-aniline
PubChem CID 82187
IUPAC Name 3-bromo-4-methylaniline
SMILES CC1=CC=C(N)C=C1Br

Specifications

Boiling Point 139°C
Quantity 5 g
UN Number 2811
Formula Weight 186.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Bromo-4-methylaniline
RTECSNumber XU4375000
TSCA No