accessibility menu, dialog, popup

UserName

H3CCH3H3COOHNNHOOOHO

CAS RN 7733-29-1

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-5-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-5-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 366.41
Molecular Formula: C18H26N2O6
InChi Key: RWQCKACYKKSOKK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NCCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)