CAS RN 77284-32-3
IUPAC Name:
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms:
(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol):
353.42
Molecular Formula:
C21H23NO4
InChi Key:
VCFCFPNRQDANPN-IBGZPJMESA-N
SMILES:
CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1
–
2
of
2
results
2
N-Fmoc-L-norleucine, 98%, Thermo Scientific Chemicals
CAS: 77284-32-3 Molecular Formula: C21H23NO4 Molecular Weight (g/mol): 353.42 MDL Number: MFCD00037537 InChI Key: VCFCFPNRQDANPN-IBGZPJMESA-N Synonym: fmoc-nle-oh,fmoc-l-norleucine,n-fmoc-l-norleucine,s-2-fmoc-amino caproic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-norleucine,fmocnleoh,fmoc-nle PubChem CID: 7009636 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid SMILES: CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O