accessibility menu, dialog, popup

UserName

CAS RN 770-30-9

IUPAC Name: 4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Synonyms: 4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Molecular Weight (g/mol): 166.2
Molecular Formula: C6H6N4S
InChi Key: VYDHBDPZNLVELI-UHFFFAOYSA-N
SMILES: CSC1=NC=C(C#N)C(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results