accessibility menu, dialog, popup

UserName

FFFHSF

CAS RN 769-40-4

IUPAC Name: 2,3,5,6-tetrafluorobenzene-1-thiol
Synonyms: 2,3,5,6-tetrafluorobenzenethiol
Molecular Weight (g/mol): 182.14
Molecular Formula: C6H2F4S
InChi Key: IGOGJHYWSOZGAE-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(S)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)