missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorothiophenol, 98%

Catalog No. AAB2348314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2348314 25 g
AAB2348306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2348314 Supplier Thermo Scientific Chemicals Supplier No. B2348314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 769-40-4 | C6H2F4S | 182.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 769-40-4
Molecular Formula C6H2F4S
Molecular Weight (g/mol) 182.136
MDL Number MFCD00004829
InChI Key IGOGJHYWSOZGAE-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
PubChem CID 69859
IUPAC Name 2,3,5,6-tetrafluorobenzenethiol
SMILES C1=C(C(=C(C(=C1F)F)S)F)F

Specifications

Density 1.53
Boiling Point 152°C to 153°C
Flash Point 48°C (119°F)
Odor Stench
Refractive Index 1.485
Quantity 25 g
UN Number UN1992
Sensitivity Air Sensitive
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorothiophenol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 212-210-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only