missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorobenzenethiol 95.0+%, TCI America™

Catalog No. T10445G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10445G Supplier TCI America Supplier No. T10445G

Chemical Identifiers

CAS 769-40-4
Molecular Formula C6H2F4S
Molecular Weight (g/mol) 182.136
MDL Number MFCD00004829
InChI Key IGOGJHYWSOZGAE-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
PubChem CID 69859
IUPAC Name 2,3,5,6-tetrafluorobenzenethiol
SMILES C1=C(C(=C(C(=C1F)F)S)F)F

Specifications

Color Yellow
Boiling Point 147°C
Quantity 5 g
UN Number 1993
Formula Weight 182.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5,6-Tetrafluorobenzenethiol