accessibility menu, dialog, popup

UserName

PP

CAS RN 7688-25-7

IUPAC Name: [4-(diphenylphosphanyl)butyl]diphenylphosphane
Synonyms: [4-(diphenylphosphanyl)butyl]diphenylphosphane
Molecular Weight (g/mol): 426.48
Molecular Formula: C28H28P2
InChi Key: BCJVBDBJSMFBRW-UHFFFAOYSA-N
SMILES: C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)