CAS RN 76824-35-6
IUPAC Name:
3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
Synonyms:
famotidine famotidina pepcid famotidinum 法莫替丁
Molecular Weight (g/mol):
337.44
Molecular Formula:
C8H15N7O2S3
InChi Key:
XUFQPHANEAPEMJ-UHFFFAOYSA-N
SMILES:
NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1
1
–
3
of
3
results
2
Famotidine 98.0+%, TCI America™
CAS: 76824-35-6 Molecular Formula: C8H15N7O2S3 Molecular Weight (g/mol): 337.435 InChI Key: XUFQPHANEAPEMJ-UHFFFAOYSA-N Synonym: famotidine,pepcid ac,apogastine,amfamox,antodine,bestidine,digervin,dispromil,famodil,gastridin PubChem CID: 5702160 ChEBI: CHEBI:4975 IUPAC Name: 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide SMILES: C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N
3
Promotions Available
Medisca Famotidine USP
Medisca Famotidine USP
Famotidine USP is a white to pale yellowish-white, crystalline powder.