accessibility menu, dialog, popup

UserName

HOBOHF

CAS RN 768-35-4

IUPAC Name: (3-fluorophenyl)boronic acid
Synonyms: 3-fluorophenylboronic acid
Molecular Weight (g/mol): 139.92
Molecular Formula: C6H6BFO2
InChi Key: KNXQDJCZSVHEIW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(F)=C1
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (2)
  • (3)
  • (4)