CAS RN 76721-89-6
IUPAC Name:
2-(2-benzyl-3-sulfanylpropanamido)acetic acid
Synonyms:
thiorphan
Molecular Weight (g/mol):
253.32
Molecular Formula:
C12H15NO3S
InChi Key:
LJJKNPQAGWVLDQ-UHFFFAOYNA-N
SMILES:
OC(=O)CNC(=O)C(CS)CC1=CC=CC=C1
1
–
1
of
1
results
1
DL-Thiorphan
CAS: 76721-89-6 Molecular Formula: C12H15NO3S Molecular Weight (g/mol): 253.32 MDL Number: MFCD00058435 InChI Key: LJJKNPQAGWVLDQ-UHFFFAOYNA-N Synonym: thiorphan,dl-thiorphan,dl-3-mercapto-2-benzylpropanoylglycine,n-2-benzyl-3-sulfanylpropanoyl glycine,chembl10247,glycine, n-2-mercaptomethyl-1-oxo-3-phenylpropyl-, +-,2-2-benzyl-3-sulfanylpropanoyl amino acetic acid,2-2-benzyl-3-mercaptopropanamido acetic acid,n-rs-2-benzyl-3-mercaptopropanoyl-glycine,tiorphan PubChem CID: 3132 ChEBI: CHEBI:9568 IUPAC Name: 2-(2-benzyl-3-sulfanylpropanamido)acetic acid SMILES: OC(=O)CNC(=O)C(CS)CC1=CC=CC=C1