accessibility menu, dialog, popup

UserName

CH3OSSOH3COH3COCH3

CAS RN 76505-71-0

IUPAC Name: 2-[(2,2-diethoxyethyl)disulfanyl]-1,1-diethoxyethane
Synonyms: 2-[(2,2-diethoxyethyl)disulfanyl]-1,1-diethoxyethane
Molecular Weight (g/mol): 298.46
Molecular Formula: C12H26O4S2
InChi Key: XJIKEAQNTPCOLU-UHFFFAOYSA-N
SMILES: CCOC(CSSCC(OCC)OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results