missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.1N in Isopropanol (Certified), Fisher Chemical

Catalog No. ST111500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
ST111500 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST111500 Supplier Thermo Fisher Scientific Supplier No. ST111 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7647-01-0 , 67-63-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 2
Melting Point -88.9°C
Boiling Point 82.8°C
Concentration 0.0995 to 0.1005N
Density 0.8g/cm³
Identification Pass Test
Packaging Amber Glass
Traceability to NIST LOT 84L Potassium acid phthalate
Vapor Pressure 33mmHg at 20°C
Quantity 500 mL
Physical Form Liquid
Grade Certified
Concentration or Composition (by Analyte or Components) 0.1N in Isopropanol
Chemical Name or Material Hydrochloric Acid Solution, 0.1N, In isopropanol
Show More Show Less