missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 2N (Certified), Fisher Chemical

Catalog No. SA431500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA431500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA431500 Supplier Thermo Fisher Scientific Supplier No. FLSA431500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Chemical Identifiers

CAS 7647-01-0 , 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

pH 0.10
Melting Point -17°C
Boiling Point 81.5°C
Concentration 1.995 to 2.005N
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 160mmHg at 20°C
Quantity 500 mL
Physical Form Liquid
Grade Certified
Concentration or Composition (by Analyte or Components) 2N
Chemical Name or Material Hydrochloric Acid Solution, 2N
Show More Show Less