accessibility menu, dialog, popup

UserName

CAS RN 76343-94-7

H3C(S)(R)(R)(R)HOO(R)SONHOOCH3(Z)(Z)

CAS RN 76343-94-7

IUPAC Name: (4R)-4-[(1R,4Z,8Z,10S,13R,15R)-15-hydroxy-5,10-dimethyl-3-oxo-2,14-dioxabicyclo[11.3.1]heptadeca-4,8-dien-15-yl]-1,3-thiazolidin-2-one
Synonyms: latrunculin B
Molecular Weight (g/mol): 395.51
Molecular Formula: C20H29NO5S
InChi Key: NSHPHXHGRHSMIK-JRIKCGFMSA-N
SMILES: C[C@H]1CC[C@@H]2C[C@H](C[C@@](O)(O2)[C@@H]2CSC(=O)N2)OC(=O)\C=C(C)/CC\C=C/1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results